2-Methoxy-5-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (6-Methoxypyridin-3-yl)boronic acid

BB-2329-1G BB-2329-5G BB-2329-25G
COBBBB-2329-1GEA 25.5 EUR
COBBBB-2329-1G COBBBB-2329-5G COBBBB-2329-25G
2-Methoxy-5-pyridinboronsäure
2-Methoxy-5-pyridinboronsäure
Formel: C₆H₈BNO₃
Molecular Weight: 152.95 g/mol
MDL: MFCD02093044
CAS-Nummer: 163105-89-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR