2-Methoxy-5-pyridinboronsäure
Lieferant: FLUOROCHEM
Synonyme:
(6-Methoxypyridin-3-yl)boronic acid
011048-5G
011048-1G
FLUO011048-5GEA
45.8
EUR
FLUO011048-5G
FLUO011048-1G
2-Methoxy-5-pyridinboronsäure
2-Methoxy-5-pyridinboronsäure
Formel:
C₆H₈BNO₃ Molecular Weight: 152.95 g/mol |
MDL:
MFCD02093044 CAS-Nummer: 163105-89-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...