2-Methoxy-5-pyridinboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (6-Methoxypyridin-3-yl)boronic acid

M-Clarity™ Qualitätsstufe = MQ400

637610-1G 637610-5G
SIAL637610-1GEA 26 EUR
SIAL637610-1G SIAL637610-5G
2-Methoxy-5-pyridinboronsäure, Sigma-Aldrich®
2-Methoxy-5-pyridinboronsäure
Formel: C₆H₈BNO₃
Molecular Weight: 152.95 g/mol
MDL: MFCD02093044
CAS-Nummer: 163105-89-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR