2,6-Dimethoxy-3-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2,6-Dimethoxypyridine-3-boronic acid, 2,6-Dimethoxypyridin-3-ylboronic acid

BB-8437-1G BB-8437-5G
COBBBB-8437-1GEA 46 EUR
COBBBB-8437-1G COBBBB-8437-5G
2,6-Dimethoxy-3-pyridinboronsäure
2,6-Dimethoxy-3-pyridineboronic acid
Formel: C₇H₁₀BNO₄
Molecular Weight: 182,97 g/mol
MDL: MFCD03788239
CAS-Nummer: 221006-70-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR