2-(Ethylthio)-5-pyridinboronsäure
Lieferant: COMBI-BLOCKS
BB-6092-1G
BB-6092-5G
COBBBB-6092-1GEA
0
EUR
COBBBB-6092-1G
COBBBB-6092-5G
2-(Ethylthio)-5-pyridinboronsäure
Formel:
C₇H₁₀BNO₂S |
MDL:
MFCD13195658 CAS-Nummer: 1217501-38-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...