2-(Methylthio)-5-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 6-(Methylthio)pyridine-3-boronic acid, 2-Methylthiopyridine-5-boronic acid, 6-(Methylthio)pyridin-3-ylboronic acid

BB-8436-5G BB-8436-1G
COBBBB-8436-5GEA 796 EUR
COBBBB-8436-5G COBBBB-8436-1G
2-(Methylthio)-5-pyridinboronsäure
2-(Methylthio)-5-pyridinboronsäure
Formel: C₆H₈BNO₂S
Molecular Weight: 169.01 g/mol
MDL: MFCD03788238
CAS-Nummer: 321438-86-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR