Dimethyl-2,5-pyridindicarboxylat

Lieferant: Thermo Scientific

Synonyme: Dimethyl pyridine-2,5-dicarboxylate

RF02989.50 RF02989.1 RF02989EA
MAYBRF02989.50EA 0 EUR
MAYBRF02989.50 MAYBRF02989.1 MAYBRF02989EA
Dimethyl-2,5-pyridindicarboxylat
Dimethyl-2,5-pyridindicarboxylat
Formel: C₉H₉NO₄
Molecular Weight: 195.17 g/mol
MDL: MFCD00034767
CAS-Nummer: 881-86-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR