Dimethyl-2,6-pyridindicarboxylat

Lieferant: COMBI-BLOCKS

Synonyme: Dimethyl pyridine-2,6-dicarboxylate

OS-7436-25G OS-7436-5G OS-7436-100G
COBBOS-7436-25GEA 75.9 EUR
COBBOS-7436-25G COBBOS-7436-5G COBBOS-7436-100G
Dimethyl-2,6-pyridindicarboxylat
Dimethyl-2,6-pyridindicarboxylat
Formel: C₉H₉NO₄
Molecular Weight: 195.17 g/mol
MDL: MFCD00134493
CAS-Nummer: 5453-67-8
EINECS: 226-697-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR