Dimethyl-2,6-pyridindicarboxylat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Dimethyl pyridine-2,6-dicarboxylate

379336-50G 379336-10G
SIAL379336-50GEA 150 EUR
SIAL379336-50G SIAL379336-10G
Dimethyl-2,6-pyridindicarboxylat, Sigma-Aldrich®
Dimethyl-2,6-pyridindicarboxylat
Formel: C₉H₉NO₄
Molecular Weight: 195,17 g/mol
MDL: MFCD00134493
CAS-Nummer: 5453-67-8
EINECS: 226-697-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR