2-Hydroxy-5-nitronicotinsäure

Lieferant: COMBI-BLOCKS

CA-4086-5G CA-4086-25G CA-4086-1G
COBBCA-4086-5GEA 0 EUR
COBBCA-4086-5G COBBCA-4086-25G COBBCA-4086-1G
2-Hydroxy-5-nitronicotinsäure
2-Hydroxy-5-nitronicotinsäure
Formel: C₆H₄N₂O₅
MDL: MFCD05669267
CAS-Nummer: 6854-07-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR