2-(2,2,2-Trifluorethoxy)-3-pyridinboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(2-(2,2,2-Trifluoroethoxy)pyridin-3-yl)boronic acid
BB-8713-1G
BB-8713-250M
COBBBB-8713-1GEA
692
EUR
COBBBB-8713-1G
COBBBB-8713-250M
2-(2,2,2-Trifluorethoxy)-3-pyridinboronsäure
2-(2,2,2-Trifluoroethoxy)-3-pyridineboronic acid
Formel:
C₇H₇BF₃NO₃ Molecular Weight: 220,9400 g/mol |
MDL:
MFCD07374887 CAS-Nummer: 1218790-79-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...