2-Methyl-3-pyridinboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
2-Methylpyridine-3-boronic acid pinacol ester, 2-Methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
PN-3922-5G
PN-3922-1G
COBBPN-3922-5GEA
560
EUR
COBBPN-3922-5G
COBBPN-3922-1G
2-Methyl-3-pyridinboronsäurepinakolester
2-Methyl-3-pyridinboronsäurepinakolester
Formel:
C₁₂H₁₈BNO₂ Molecular Weight: 219.0878 g/mol |
MDL:
MFCD08669594 CAS-Nummer: 1012084-56-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...