2-Methyl-4-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-Methylpyridine-4-boronic acid Pinacol ester, 2-(2-Methyl-4-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

PN-8898-5G PN-8898-1G
COBBPN-8898-5GEA 352 EUR
COBBPN-8898-5G COBBPN-8898-1G
2-Methyl-4-pyridinboronsäurepinakolester
2-Methyl-4-pyridinboronsäurepinakolester
Formel: C₁₂H₁₈BNO₂
Molecular Weight: 219.09 g/mol
MDL: MFCD08061590
CAS-Nummer: 660867-80-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR