2-Methyl-5-pyridinboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
2-Methylpyridine-5-boronic acid pinacol ester, 6-Methylpyridine-3-boronic acid pinacol ester, 2-(6-Methyl-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 6-Methyl-3-pyridylboronic acid Pinacol ester
PN-5068-5G
PN-5068-1G
COBBPN-5068-5GEA
158
EUR
COBBPN-5068-5G
COBBPN-5068-1G
2-Methyl-5-pyridinboronsäurepinakolester
2-Methyl-5-pyridinboronsäurepinakolester
Formel:
C₁₂H₁₈BNO₂ Molecular Weight: 219.0878 g/mol |
MDL:
MFCD08669608 CAS-Nummer: 610768-32-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...