2-Methyl-5-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (6-Methylpyridin-3-yl)boronic acid

BB-5068-5G BB-5068-1G
COBBBB-5068-5GEA 259 EUR
COBBBB-5068-5G COBBBB-5068-1G
2-Methyl-5-pyridinboronsäure
2-Methyl-5-pyridinboronsäure
Formel: C₆H₈BNO₂
Molecular Weight: 136,94 g/mol
MDL: MFCD06801736
CAS-Nummer: 659742-21-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR