2-Chlor-6-(ethoxycarbonyl)-4-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

PN-5854-1G PN-5854-250M
COBBPN-5854-1GEA 1370 EUR
COBBPN-5854-1G COBBPN-5854-250M
2-Chlor-6-(ethoxycarbonyl)-4-pyridinboronsäurepinakolester
2-Chlor-6-(ethoxycarbonyl)-4-pyridinboronsäurepinakolester
Formel: C₁₄H₁₉BClNO₄
MDL: MFCD11617858
CAS-Nummer: 741709-70-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR