2-Chlor-5-methyl-3-pyridinboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
2-CHLORO-5-METHYLPYRIDINE-3-BORONIC ACID 98
BB-3511-5G
BB-3511-1G
COBBBB-3511-5GEA
459
EUR
COBBBB-3511-5G
COBBBB-3511-1G
2-Chlor-5-methyl-3-pyridinboronsäure
2-Chloro-5-methyl-3-pyridineboronic acid
Formel:
C₆H₇BClNO₂ Molecular Weight: 171.3900 g/mol |
MDL:
MFCD04112563 CAS-Nummer: 913835-86-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...