2-Chlor-6-isopropyl-3-pyridinboronsäure

Lieferant: COMBI-BLOCKS

BB-9049-5G BB-9049-1G
COBBBB-9049-5GEA 0 EUR
COBBBB-9049-5G COBBBB-9049-1G
2-Chlor-6-isopropyl-3-pyridinboronsäure
2-Chlor-6-isopropyl-3-pyridinboronsäure
Formel: C₈H₁₁BClNO₂
MDL: MFCD08274472
CAS-Nummer: 1003043-37-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR