2-Chlor-3-methyl-5-pyridinboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
2-Chloro-3-methylpyridine-5-boronic acid, 6-Chloro-5-methylpyridine-3-boronic acid, (6-Chloro-5-methylpyridin-3-yl)boronic acid
BB-8415-5G
BB-8415-1G
COBBBB-8415-5GEA
205
EUR
COBBBB-8415-5G
COBBBB-8415-1G
2-Chlor-3-methyl-5-pyridinboronsäure
2-Chlor-3-methyl-5-pyridinboronsäure
Formel:
C₆H₇BClNO₂ Molecular Weight: 171.39 g/mol |
MDL:
MFCD03411564 CAS-Nummer: 1003043-40-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...