4-Methylnicotinsäure Hydrochlorid

Lieferant: Thermo Scientific

SB01942DA SB01942EA
MAYBSB01942DAEA 0 EUR
MAYBSB01942DA MAYBSB01942EA
4-Methylnicotinsäure Hydrochlorid
4-Methylnicotinsäure Hydrochlorid
Formel: C₇H₈ClNO₂
Molecular Weight: 173,60 g/mol
MDL: MFCD00832700
CAS-Nummer: 94015-05-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR