2-Fluor-5-methyl-3-pyridinboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
2-Fluoro-5-methylpyridine-3-boronic acid pinacol ester, 2-Fluoro-5-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
PN-8901-5G
PN-8901-1G
COBBPN-8901-5GEA
692
EUR
COBBPN-8901-5G
COBBPN-8901-1G
2-Fluor-5-methyl-3-pyridinboronsäurepinakolester
2-Fluor-5-methyl-3-pyridinboronsäurepinakolester
Formel:
C₁₂H₁₇BFNO₂ Molecular Weight: 237,0783 g/mol |
MDL:
MFCD08063092 CAS-Nummer: 1073371-96-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...