2-Fluor-3-methyl-5-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-Fluoro-3-methylpyridine-5-boronic acid, pinacol ester, 2-Fluoro-3-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

PN-8413-1G PN-8413-250M
COBBPN-8413-1GEA 595 EUR
COBBPN-8413-1G COBBPN-8413-250M
2-Fluor-3-methyl-5-pyridinboronsäurepinakolester
2-Fluor-3-methyl-5-pyridinboronsäurepinakolester
Formel: C₁₂H₁₇BFNO₂
Molecular Weight: 237.800 g/mol
MDL: MFCD11855990
CAS-Nummer: 1150561-71-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR