2-(Trifluormethyl)-3-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-(Trifluoromethyl)pyridin-3-yl)boronic acid

BB-8584-250M BB-8584-100M
COBBBB-8584-250MEA 154 EUR
COBBBB-8584-250M COBBBB-8584-100M
2-(Trifluormethyl)-3-pyridinboronsäure
2-(Trifluoromethyl)-3-pyridineboronic acid
Formel: C₆H₅BF₃NO₂
Molecular Weight: 190,92 g/mol
MDL: MFCD09271791
CAS-Nummer: 947533-39-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR