2-Fluor-4-methyl-3-pyridinboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
2-Fluoro-4-methylpyridine-3-boronic acid, (2-Fluoro-4-methylpyridin-3-yl)boronic acid
BB-8523-1G
BB-8523-100M
BB-8523-250M
COBBBB-8523-1GEA
1630
EUR
COBBBB-8523-1G
COBBBB-8523-100M
COBBBB-8523-250M
2-Fluor-4-methyl-3-pyridinboronsäure
2-Fluoro-4-methyl-3-pyridineboronic acid
Formel:
C₆H₇BFNO₂ Molecular Weight: 154.940 g/mol |
MDL:
MFCD07374980 CAS-Nummer: 1029654-30-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...