5-(Trifluormethyl)-2-pyridinboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(5-(Trifluoromethyl)pyridin-2-yl)boronic acid
BB-0102-1G
BB-0102-250M
COBBBB-0102-1GEA
596
EUR
COBBBB-0102-1G
COBBBB-0102-250M
5-(Trifluormethyl)-2-pyridinboronsäure
5-(Trifluoromethyl)-2-pyridineboronic acid
Formel:
C₆H₅BF₃NO₂ Molecular Weight: 190.9156 g/mol |
MDL:
MFCD06227445 CAS-Nummer: 1162257-58-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...