3-(Trifluormethyl)-5-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (5-(Trifluoromethyl)pyridin-3-yl)boronic acid, (5-Trifluoromethylpyridin-3-yl)boronic acid

BB-5273-1G BB-5273-250M
COBBBB-5273-1GEA 449 EUR
COBBBB-5273-1G COBBBB-5273-250M
3-(Trifluormethyl)-5-pyridinboronsäure
3-(Trifluormethyl)-5-pyridinboronsäure
Formel: C₆H₅BF₃NO₂
Molecular Weight: 190.9156 g/mol
MDL: MFCD09952041
CAS-Nummer: 947533-51-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR