2-Ethoxy-5-(trifluormethyl)-3-pyridinboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(2-Ethoxy-5-(trifluoromethyl)pyridin-3-yl)boronic acid
BB-6236-1G
BB-6236-5G
COBBBB-6236-1GEA
175
EUR
COBBBB-6236-1G
COBBBB-6236-5G
2-Ethoxy-5-(trifluormethyl)-3-pyridinboronsäure
2-Ethoxy-5-(trifluoromethyl)-3-pyridineboronic acid
Formel:
C₈H₉BF₃NO₃ Molecular Weight: 234,9682 g/mol |
MDL:
MFCD15143470 CAS-Nummer: 1218790-66-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...