2-(Methylthio)-5-(trifluormethyl)-3-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-(Methylthio)-5-(trifluoromethyl)pyridin-3-yl)boronic acid

BB-6384-1G BB-6384-250M
COBBBB-6384-1GEA 157 EUR
COBBBB-6384-1G COBBBB-6384-250M
2-(Methylthio)-5-(trifluormethyl)-3-pyridinboronsäure
2-(Methylthio)-5-(trifluoromethyl)-3-pyridineboronic acid
Formel: C₇H₇BF₃NO₂S
Molecular Weight: 237.100 g/mol
MDL: MFCD16295069
CAS-Nummer: 1256346-08-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR