2-Amino-4,6-dimethylnicotinsäure

Lieferant: BIONET RESEARCH

9C-087.500 9C-087.1 9C-087.5 9C-087.10
BIOT9C-087.500EA 0 EUR
BIOT9C-087.500 BIOT9C-087.1 BIOT9C-087.5 BIOT9C-087.10
2-Amino-4,6-dimethylnicotinsäure
2-Amino-4,6-dimethylnicotinsäure
Formel: C₈H₁₀N₂O₂
MDL: MFCD00138571
CAS-Nummer: 106837-89-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR