2-Benzyloxy-3-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-(Benzyloxy)pyridine-3-boronic acid, (2-(Benzyloxy)pyridin-3-yl)boronic acid

BB-8506-1G BB-8506-250M
COBBBB-8506-1GEA 462 EUR
COBBBB-8506-1G COBBBB-8506-250M
2-Benzyloxy-3-pyridinboronsäure
2-Benzyloxy-3-pyridinboronsäure
Formel: C₁₂H₁₂BNO₃
Molecular Weight: 229,04 g/mol
MDL: MFCD06798264
CAS-Nummer: 1072952-41-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR