2-Benzyloxy-4-pyridinboronsäure

Lieferant: COMBI-BLOCKS

FA-3254-1G FA-3254-250M
COBBFA-3254-1GEA 983 EUR
COBBFA-3254-1G COBBFA-3254-250M
2-Benzyloxy-4-pyridinboronsäure
2-Benzyloxy-4-pyridinboronsäure
Formel: C₁₂H₁₂BNO₃
MDL: MFCD12407141
CAS-Nummer: 1256355-62-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR