2-Benzyloxy-5-pyridinboronsäure

Lieferant: COMBI-BLOCKS

BB-5175-5G BB-5175-1G
COBBBB-5175-5GEA 0 EUR
COBBBB-5175-5G COBBBB-5175-1G
2-Benzyloxy-5-pyridinboronsäure
2-Benzyloxy-5-pyridinboronsäure
Formel: C₁₂H₁₂BNO₃
MDL: MFCD06798262
CAS-Nummer: 929250-35-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR