2-Fluor-6-phenyl-3-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Fluoro-6-phenylpyridine-3-boronic acid

FA-1409-250M FA-1409-100M
COBBFA-1409-250MEA 497 EUR
COBBFA-1409-250M COBBFA-1409-100M
2-Fluor-6-phenyl-3-pyridinboronsäure
2-Fluoro-6-phenyl-3-pyridineboronic acid
Formel: C₁₁H₉BFNO₂
MDL: MFCD07437944
CAS-Nummer: 1029654-19-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR