Bismut(III)-2-ethylhexanoat ∼92% in 2-Ethylhexansäure
Lieferant: Thermo Scientific
Achtung
Synonyme: Bismuth 2-ethylhexanoate
45500.06 45500.14 45500.22
45500.06EA 29.9 EUR
45500.06 45500.14 45500.22
Bismut(III)-2-ethylhexanoat ∼92% in 2-Ethylhexansäure
Bismut(III)-2-ethylhexanoat
Purity: 92% CAS No.: 67874-71-9 Molecular Formula: C24H45BiO6 Formula Weight: 638.61 Physical Form: Liquid
Formel: C₂₄H₄₅BiO₆
Molecular Weight: 638,598 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00043803
CAS-Nummer: 67874-71-9
EINECS: 267-499-7