Methylcyclopentadienylmangantricarbonyl, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

317632-1G 317632-50G
SIAL317632-1GEA 22.9 EUR
SIAL317632-1G SIAL317632-50G
Methylcyclopentadienylmangantricarbonyl, Sigma-Aldrich®
Methylcyclopentadienylmangantricarbonyl
Formel: C₉H₇MnO₃
Molecular Weight: 218,09 g/mol
MDL: MFCD00001426
CAS-Nummer: 12108-13-3
EINECS: 235-166-5
UN: 3281
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR