1,2-Benzisoxazol-3-amin
Lieferant: Thermo Scientific
MO00344
MO00344CB
MO00344DA
MAYBMO00344EA
0
EUR
MAYBMO00344
MAYBMO00344CB
MAYBMO00344DA
1,2-Benzisoxazol-3-amin
1,2-Benzisoxazol-3-amin
Formel:
C₇H₆N₂O |
MDL:
MFCD03407353 CAS-Nummer: 36216-80-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...