9-Ethinyl-9-fluorenol ≥98%
Lieferant: Thermo Scientific
Synonyme: 9-Ethynyl-9H-fluoren-9-ol
L09717.03 L09717.06
L09717.03EA 33.9 EUR
L09717.03 L09717.06
9-Ethinyl-9-fluorenol ≥98%
9-Ethinyl-9-fluorenol
Purity: 98% CAS No.: 13461-74-0 Molecular Formula: C15H10O Formula Weight: 206.25 Melting Point: 109-110° Beilstein Reference: 2104685
Formel: C₁₅H₁₀O
Molecular Weight: 206,25 g/mol
Schmelzpunkt: 109…110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00041555
CAS-Nummer: 13461-74-0