3-(2,6-Dichlorphenyl)-5-methyl-4-isoxazolcarbonsäure
Lieferant: Thermo Scientific
SB00572.1
SB00572EA
SB00572EE
MAYBSB00572.1EA
0
EUR
MAYBSB00572.1
MAYBSB00572EA
MAYBSB00572EE
3-(2,6-Dichlorphenyl)-5-methyl-4-isoxazolcarbonsäure
3-(2,6-Dichlorophenyl)-5-methylisoxazole-4-carboxylic acid
Formel:
C₁₁H₇Cl₂NO₃ Molecular Weight: 272.09 g/mol |
MDL:
MFCD00051658 CAS-Nummer: 3919-76-4 EINECS: 223-487-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...