3-(2-Chlor-6-fluorphenyl)-5-methylisoxazol-4-carbonitril

Lieferant: FLUOROCHEM

008238-1G 008238-5G
FLUO008238-1GEA 125 EUR
FLUO008238-1G FLUO008238-5G
3-(2-Chlor-6-fluorphenyl)-5-methylisoxazol-4-carbonitril
3-(2-Chlor-6-fluorphenyl)-5-methylisoxazol-4-carbonitril
Formel: C₁₁H₆ClFN₂O
MDL: MFCD00052551
CAS-Nummer: 175204-41-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR