3-(2-Chlor-6-fluorphenyl)-5-methyl-4-isoxazolcarbonsäureamid
Lieferant: Thermo Scientific
RF00732DA
RF00732EA
MAYBRF00732DAEA
0
EUR
MAYBRF00732DA
MAYBRF00732EA
3-(2-Chlor-6-fluorphenyl)-5-methyl-4-isoxazolcarbonsäureamid
3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carboxamide
Formel:
C₁₁H₈ClFN₂O₂ |
MDL:
MFCD00173954 CAS-Nummer: 4415-11-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...