3-(2-Chlor-6-fluorphenyl)-5-methyl-4-isoxazolcarbonsäure

Lieferant: Thermo Scientific

SB01884DA SB01884EA SB01884.50
MAYBSB01884DAEA 0 EUR
MAYBSB01884DA MAYBSB01884EA MAYBSB01884.50
3-(2-Chlor-6-fluorphenyl)-5-methyl-4-isoxazolcarbonsäure
3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carboxylic acid
Formel: C₁₁H₇ClFNO₃
Molecular Weight: 255.63 g/mol
MDL: MFCD00055659
CAS-Nummer: 3919-74-2
EINECS: 223-486-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR