3-(4-Fluorphenyl)-5-methyl-4-isoxazolcarbonsäure ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3-(4-Fluorophenyl)-5-methylisoxazol-4-carboxylic acid
H32090.03
H32090.06
H32090.03EA
59.4
EUR
H32090.03
H32090.06
3-(4-Fluorphenyl)-5-methyl-4-isoxazolcarbonsäure ≥97%
3-(4-Fluorophenyl)-5-methylisoxazole-4-carboxylic acid
Purity: 97%
CAS No.: 1736-21-6
Molecular Formula: C11H8FNO3
Formula Weight: 221.19
Storage Temperature: 2°C to 8°C
Melting Point: 199-201°
MDL No.: MFCD03407356
Formel:
C₁₁H₈FNO₃ Molecular Weight: 221.19 g/mol Schmelzpunkt: 199…201 °C |
MDL:
MFCD03407356 CAS-Nummer: 1736-21-6 |