3,6-Dimethoxy-2-nitrobenzaldehyd
Lieferant: BIONET RESEARCH
7W-0313.500
7W-0313.1
7W-0313.5
7W-0313.10
BIOT7W-0313.500EA
0
EUR
BIOT7W-0313.500
BIOT7W-0313.1
BIOT7W-0313.5
BIOT7W-0313.10
3,6-Dimethoxy-2-nitrobenzaldehyd
3,6-Dimethoxy-2-nitrobenzaldehyd
Formel:
C₉H₉NO₅ |
MDL:
MFCD00091893 CAS-Nummer: 1206-55-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...