2-Ethoxy-5-(trifluormethyl)anilin

Lieferant: COMBI-BLOCKS

HC-6983-1G HC-6983-5G
COBBHC-6983-1GEA 0 EUR
COBBHC-6983-1G COBBHC-6983-5G
2-Ethoxy-5-(trifluormethyl)anilin
2-Ethoxy-5-(trifluormethyl)anilin
Formel: C₉H₁₀F₃NO
MDL: MFCD01422864
CAS-Nummer: 2713-73-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR