Isobutyramid 99

Lieferant: Thermo Fisher Scientific
Achtung

B21641.14 B21641.22 B21641.36
B21641.14EA 56.9 EUR
B21641.14 B21641.22 B21641.36
Isobutyramid 99
Isobutyramid

Purity: 99% CAS No.: 563-83-7 Molecular Formula: (CH3)2CHCONH2 Formula Weight: 87.12 Melting Point: 127-129° Boiling Point: 216-220° MDL No.: MFCD00008019

Formel: C₄H₉NO
Molecular Weight: 87.12 g/mol
Siedepunkt: 216…220 °C
Schmelzpunkt: 127…129 °C
Dichte: 1.013
Lagertemperatur: Raumtemperatur
MDL: MFCD00008019
CAS-Nummer: 563-83-7
EINECS: 209-265-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR