Isobutyramid, Sigma-Aldrich®

Lieferant: Merck


MQ100

144436-100G 144436-25G
SIAL144436-100GEA 176 EUR
SIAL144436-100G SIAL144436-25G
Isobutyramid, Sigma-Aldrich®
Isobutyramid
Formel: C₄H₉NO
Molecular Weight: 87.12 g/mol
MDL: MFCD00008019
CAS-Nummer: 563-83-7
EINECS: 209-265-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR