2,4-Bis(trifluormethyl)toluol

Lieferant: FLUOROCHEM

006045-5G 006045-25G 006045-1G
FLUO006045-5GEA 533 EUR
FLUO006045-5G FLUO006045-25G FLUO006045-1G
2,4-Bis(trifluormethyl)toluol
2,4-Bis(trifluormethyl)toluol
Formel: C₉H₆F₆
Molecular Weight: 228.134 g/mol
MDL: MFCD00792411
CAS-Nummer: 136664-96-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR