4-Hydroxynicotinsäure

Lieferant: COMBI-BLOCKS

CA-4068-5G CA-4068-1G
COBBCA-4068-5GEA 122 EUR
COBBCA-4068-5G COBBCA-4068-1G
4-Hydroxynicotinsäure
4-Hydroxynicotinsäure
Formel: C₆H₅NO₃
Molecular Weight: 139.11 g/mol
MDL: MFCD00040286
CAS-Nummer: 609-70-1
EINECS: 210-197-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR