2,5-Bis(trifluormethyl)benzaldehyd

Lieferant: CHEMPUR

FL13757.POR 013757-5G
CHMPFL13757.POREA 0 EUR
CHMPFL13757.POR FLUO013757-5G
2,5-Bis(trifluormethyl)benzaldehyd
2,5-Bis(trifluormethyl)benzaldehyd
Formel: C₉H₄F₆O
Molecular Weight: 242,12 g/mol
MDL: MFCD00674087
CAS-Nummer: 395-64-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR