(1S,2S)-trans-2-Aminocyclopentanol Hydrochlorid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
trans-(1S,2S)-2-Aminocyclopentanol Hydrochloride, (1S,2S)-2-Aminocyclopentanol hydrochloride, (1S,2S)-trans-2-Aminocyclopentanol hydrochloride
M-Clarity™ Qualitätsstufe = MQ100
06782-1G-F
06782-5G-F
SIAL06782-1G-FEA
250
EUR
SIAL06782-1G-F
SIAL06782-5G-F
(1S,2S)-trans-2-Aminocyclopentanol Hydrochlorid, Sigma-Aldrich®
(1S,2S)-trans-2-Aminocyclopentanol Hydrochlorid
Formel:
C₅H₁₂ClNO Molecular Weight: 137.6079 g/mol |
MDL:
MFCD07370092 CAS-Nummer: 68327-04-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...